Anti-CD4 Rat Monoclonal Antibody (BG Violet 450) [clone: RM4-5]

Supplier: BIOGEMS INTERNATIONAL INC.
06122-40-100
75844-026EA 232.14 USD
75844-026
Anti-CD4 Rat Monoclonal Antibody (BG Violet 450) [clone: RM4-5]
Antibodies
The RM4-5 monoclonal antibody specifically reacts with mouse CD4, also known as L3T4, a 55 kDa differentiation antigen expressed by the majority of thymocytes, subpopulations of mature T cells (like major histocompatibility complex class II-restricted T lymphocytes), a subset of natural killer T cells, and on pluripotent hematopoietic stem cells. CD4 binds to the major histocompatibility complex class II (MHC class II) and enhances T lymphocyte development and mature T cells functions. In T lymphocytes, CD4 binds to the cytoplasmic tail of enzyme tyrosine kinase (p56lck).Binding of RM4-5 is blocked by the anti-mouse CD4 clone GK1.5.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RM4-5
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR