Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RPA-T4]

Supplier: Biogems
06121-50-100
75843-996EA 116.13 USD
75843-996
Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RPA-T4]
Antibodies
The RPA-T4 monoclonal antibody specifically binds to the CD4 receptor for the human immunodeficiency virus (HIV). CD4 is a 59 kDa single-chain transmembrane glycoprotein that expressed on the surface of most of the thymocytes, T-helper cells, and in low levels on monocytes and macrophages. CD4 is a co-receptor in the antigen-induced T cell activation, together with the MHC class II. The RPA-T4 antibody is capable of blocking HIV binding and inhibiting syncytium formation, by binding to the D1 domain of the CD4 antigen. The OKT4 and the RPA-T4 monoclonal antibodies recognize different epitopes of CD4 and they do not exhibit cross-block binding.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR