CD4 MAB mg
Supplier: Biogems
CD4, Monoclonal Antibody, Clone: GK1.5, Host: Rat, Species Reactivity: Mouse, Isotype: IgG2b, kappa, Conjugate: SAFIRE Purified, Formulations: Phosphate-buffered aqueous solution, less than or equal to 0.09% Sodium azide, protein/stabilizer, ph7.2, Application: Flow Cytometry, Size: mg