Anti-CD4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OKT4]

Supplier: BIOGEMS INTERNATIONAL INC.
06111-80-500
75843-950EA 806.51 USD
75843-950
Anti-CD4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OKT4]
Antibodies
The OKT4 monoclonal antibody specifically binds to the CD4 receptor for the human immunodeficiency virus (HIV). CD4 is a 59 kDa single-chain transmembrane glycoprotein that expressed on the surface of most of the thymocytes, T-helper cells, and in low levels on monocytes and macrophages. CD4 is a co-receptor in the antigen-induced T cell activation (together with the MHC class II). The OKT4 and the RPA-T4 monoclonal antibodies recognize different epitopes of CD4 and they do not exhibit cross-block binding.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: OKT4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR