Anti-CD83 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: Michel17]

Supplier: Biogems
05912-60-100
75843-928EA 570.02 USD
75843-928
Anti-CD83 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: Michel17]
Antibodies
The Michel-17 monoclonal antibody specifically reacts with mouse CD83, a 45kDA type I transmembrane glycoprotein. CD83 is expressed mainly on activated lymphocytes and mature dendritic cells and it plays a role in T cell development. The Michel-17 antibody does not induce an activation signal on dendritic or T cells.

Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: Michel17
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR