Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: SK7]

Supplier: Biogems
05141-77-100
75843-888EA 393.15 USD
75843-888
Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: SK7]
Antibodies
The SK7 monoclonal antibody specifically reacts with the ε chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains γ, δ, and ε chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction. The SK7 antibody is reported to have a mitogenic effect on the majority of peripheral blood T cells in the presence of functional monocytes.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: SK7
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR