Anti-CD3 Mouse Monoclonal Antibody (BG Violet 450) [clone: SK7]

Supplier: BIOGEMS INTERNATIONAL INC.
05141-40-25
75843-886EA 154.85 USD
75843-886
Anti-CD3 Mouse Monoclonal Antibody (BG Violet 450) [clone: SK7]
Antibodies
The SK7 monoclonal antibody specifically reacts with the ε chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains γ, δ, and ε chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction. The SK7 antibody is reported to have a mitogenic effect on the majority of peripheral blood T cells in the presence of functional monocytes.

BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: SK7
Conjugation: BG Violet 450
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR