Anti-CD3 Mouse Monoclonal Antibody (BG Violet 450) [clone: UCHT1]

Supplier: BIOGEMS INTERNATIONAL INC.
05131-40-100
75843-850EA 264.54 USD
75843-850
Anti-CD3 Mouse Monoclonal Antibody (BG Violet 450) [clone: UCHT1]
Antibodies
The UCHT1 monoclonal antibody specifically reacts with the ε chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains γ, δ, and ε chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction. Unlike HIT3a, another specific antibody of CD3, the UCHT1 antibody can stain both the surface and intracellular CD3ε. The immobilized UCHT1 can cross-link with the TCR complex, enhancing cellular activation and proliferation.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation: BG Violet 450
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR