Anti-CD3e Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)-Cy5®) [clone: 145-2C11]

Supplier: Biogems
05122-65-100
75843-824EA 206.45 USD
75843-824
Anti-CD3e Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)-Cy5®) [clone: 145-2C11]
Antibodies
The 145-2C11 monoclonal antibody reacts with mouse CD3e, the 20 kDa ε chain of the TCR complex. Together with the γ and δ subunits of CD3, the ε subunits are involved in the assembly, trafficking, and surface expression of T-cell receptor complex. CD3 is expressed on thymocytes, mature T cells, and natural killer T cells, and the ε chain enhances the antigen recognition.The 145-2C11 antibody binds to the TCR complex and, depending on the conditions, initiates T cells activation, proliferation, and apoptosis. The soluble antibody seems to block lysis of target cells by antigen-specific cytotoxic T lymphocytes. The 145-2C11 antibody does not cross-react with the rat leukocytes, and it is used as a phenotypic marker for mouse T lymphocytes.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: 145-2C11
Conjugation: PE (Phycoerythrin)-Cy5®
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR