Anti-CD42b Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HIP1]

Supplier: BIOGEMS INTERNATIONAL INC.
04511-50-100
75843-812EA 511.39 USD
75843-812
Anti-CD42b Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HIP1]
Antibodies
The HIP1 monoclonal antibody specifically binds to human CD42b, a 145kDa transmembrane protein known as the platelet glycoprotein Ib alpha chain (gpIbα). CD42 is expressed on platelets and megakaryocytes and is reported to be involved in platelet adhesion. The HIP1 antibody inhibits collage-induced aggregation and platelet to von Willebrand Factor (vWF) binding.

Type: Primary
Antigen: CD42b
Clonality: Monoclonal
Clone: HIP1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR