Anti-CD32 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 6C4 ]

Supplier: BIOGEMS INTERNATIONAL INC.
04411-80-25
75843-810EA 339.03 USD
75843-810
Anti-CD32 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 6C4 ]
Antibodies
The 6C4 monoclonal antibody specifically recognizes two isoforms of human CD32, Fc gamma RIIA and Fc gamma RIIB. From alternative mRNA splicing, 6 isoforms of CD32 have been reported. CD32 is a 40 kDA polymorphic transmembrane glycoprotein expressed on monocytes, macrophages, graulocytes, B cells, and platelets. The receptor has low affinity toward IgG and is involved in immunomodulation, phagocytosis, and the inflammation response. The 6C4 antibody in a rosette blocking assay is reported to inhibit Ig binding

Type: Primary
Antigen: CD32
Clonality: Monoclonal
Clone: 6C4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR