Anti-CD2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RM2-5]

Supplier: BIOGEMS INTERNATIONAL INC.
04112-60-100
75843-800EA 258.08 USD
75843-800
Anti-CD2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RM2-5]
Antibodies
The RM2-5 monoclonal antibody specifically binds to mouse CD2, a member of the Ig superfamily and type I transmembrane glycoprotein. CD2 is expressed on B cells, T cells, NK cells, and thymocytes and its ligand is CD48. It may be involved in immunoregulation, thymocyte maturation, and T-cell activation. The RM2-5 antibody has been reported to block CD2 mediated cell to cell adhesion.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RM2-5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b, lambda
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR