CD90 (Thy-1) MAB 100tests
Supplier: Biogems
CD90 (Thy-1), Monoclonal Antibody, Clone: 5E10, Host: Mouse, Species: Human, Isotype: IgG1, kappa, Conjugate: FITC, Formulations: Phosphate-buffered aqueous solution, less than or equal to 0.09% Sodium azide, protein/stabilizer, ph7.2, Application: FC, Size: 100tests