Anti-F4/80 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: BM8.1]

Supplier: Biogems
02922-80-100
75843-762EA 290.34 USD
75843-762
Anti-F4/80 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: BM8.1]
Antibodies
The BM8.1 monoclonal antibody specifically binds to the Mouse125 kDa F4/80 antigen, expressed by most mature macrophages. F4/80 is a transmembrane protein used as a marker of macrophages, although it is also expressed on Kupffer and Langerhans cells. The expression of F4/80 antigen is upregulated on bone marrow cells stimulated in vitro with the macrophage colony stimulating factor. The F4/80 antigen is a requirement for the induction of CD8 T cells-mediated peripheral tolerance.

Type: Primary
Antigen: F4/80
Clonality: Monoclonal
Clone: BM8.1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR