CD80 (B7-1) MAB 100ug
Supplier: Biogems
CD80 (B7-1), Monoclonal Antibody, Clone: 3H5, Host: Mouse, Species Reactivity: Rat, Isotype: IgG1, kappa, Conjugate: PE, Formulations: Phosphate-buffered aqueous solution, less than or equal to 0.09% Sodium azide, protein/stabilizer, ph7.2, Application: FC, Size: 100ug