CD80 (B7-1) MAB 100ug
Supplier: Biogems
CD80 (B7-1), Monoclonal Antibody, Clone: 16-10A1, Host: Armenian Hamster, Species: Mouse, Isotype: IgG, Conjugate: Purified, Formulations: Phosphate-buffered aqueous solution, less than or equal to 0.09% Sodium azide, protein/stabilizer, ph7.2, Application: FC, Size: 100ug