Anti-CD40 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HM40-3]

Supplier: Biogems
02512-50-50
75843-692EA 211.16 USD
75843-692
Anti-CD40 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HM40-3]
Antibodies
The HM40-3 monoclonal antibody reacts with mouse CD40, a 48kDA type I glycoprotein known as Bp50. CD40 is part of the TNFR family and expressed on a subset of T cells, B cells, dendritic cells, and macrophages. It has been shown to be involved in the protection of B cells from apoptosis, B cell differentiation, co-stimulation, and isotype class-switching. The HM40-3 antibody blocks the binding of CD40L (CD154) and stimulates B and dendritic cell proliferation.

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: HM40-3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgM, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR