Anti-TCRB Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: H57-597]

Supplier: Biogems
89612-80-25
75843-672EA 83.87 USD
75843-672
Anti-TCRB Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: H57-597]
Antibodies
The H57-597 monoclonal antibody is specific for the beta chain of the mouse T cell Receptor (TCR). The crosslinking induces activation and proliferation of T cells, as a plate-bound or a soluble H57-597, or the plate-bound antibody can induce apoptosis, based on assay conditions. The beta chain of the TCR can combine with the alpha chain of the receptor to produce the alpha-beta TCR, which is expressed by the NKT cells, by the NK1.1+ thymocytes and most of the T cells. The beta chain does not react with the gamma-delta TCR-bearing cells, expressed by a small number of T cells. This antibody can be used as a phenotypic marker for the TCR beta expressing cells or for the functional purpose of TCR-mediated cell activation or apoptosis.

Type: Primary
Antigen: TCRB
Clonality: Monoclonal
Clone: H57-597
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR