Anti-Ly-6G Rat Monoclonal Antibody (BG Violet 450) [clone: 1A8]

Supplier: BIOGEMS INTERNATIONAL INC.
83112-40-25
75843-620EA 109.69 USD
75843-620
Anti-Ly-6G Rat Monoclonal Antibody (BG Violet 450) [clone: 1A8]
Antibodies
The 1A8 monoclonal antibody specifically reacts with the 21-25 kDa glycophosphatidylinositol-anchored protein known as Ly-6G, expressed by the granulocytes from the bone marrow and periphery neutrophils. Ly-6G and Ly-6C form the Granulocyte Receptor-1 antigen (GR-1). The binding of the 1A8 antibody to the Ly-6G can be blocked by another antibody, RB6-8C5, which also recognizes Ly-6C. While 1A8 is specific for only Ly-6G, RB6-8C5 also binds to Ly-6C.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Primary
Antigen: Ly-6G
Clonality: Monoclonal
Clone: 1A8
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR