Anti-KLRG1 Golden Syrian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2F1]

Supplier: BIOGEMS INTERNATIONAL INC.
82112-60-25
75843-584EA 96.79 USD
75843-584
Anti-KLRG1 Golden Syrian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2F1]
Antibodies
The 2F1 monoclonal antibody is specific for the mouse Killer cell Lectin-like Receptor G1 (KLRG1), a homodimer consisting of two N-glycosylated subunits of 30-38 kDa, also known as MAFA (Mast cell Function-associated Antigen). The antigen contains a cytoplasmic motif similar to ITIM (the immunoreceptor tyrosine-based inhibitory motif). KLRG1 is a receptor for cadherin, a family of transmembrane glycoproteins that mediate cell adhesion, and a common marker of T cell senescence. The receptor is believed to play an important role in the innate and adaptive immune system through the regulation of leukocytes. It is expressed on lymphokine-activated killer (LAK) cells, adherent LAK (A-LAK) cells, a sub-set of natural killer (NK) cells, T cells. In NK cells, it inhibits cytokine production and cytotoxicity activity.The receptor expression was not detected on the mouse peritoneal mast cells, or bone marrow mast cells.

Type: Primary
Antigen: KLRG1
Clonality: Monoclonal
Clone: 2F1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Golden Syrian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR