Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2]

Supplier: BIOGEMS INTERNATIONAL INC.
81121-25-500
75843-524EA 296.79 USD
75843-524
Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2]
Antibodies
The MP4-25D2 monoclonal antibody specifically reacts with human Interleukin-4 (IL-4), expressed by the activated T cells and mast cells. IL-4 is species-specific and stimulates the proliferation and differentiation of B lymphocytes. It upregulates the expression of IgE receptors and class II MHC antigen. The MP4-25D2 is a neutralizing antibody and is reported to be cross-reactive with rhesus monkeys.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR