Anti-IL-4 Rat Monoclonal Antibody [clone: 11B11]

Supplier: Biogems
81112-25-100
75843-510EA 238.73 USD
75843-510
Anti-IL-4 Rat Monoclonal Antibody [clone: 11B11]
Antibodies
The 11B11 monoclonal antibody specifically reacts with mouse 14 kDa cytokine IL-4, expressed by the activated NKT cells and mast cells. IL-4 is species-specific and stimulates the proliferation and differentiation of B lymphocytes. It upregulates the expression of IgE receptors and class II MHC antigen.The 11B11 antibody neutralizes the IL-4 cytokine.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: 11B11
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR