IL-4 MAB 100 ug
Supplier: Biogems
IL-4, monoclonal Antibody, Clone: 11B11, Host: Rat, Species reactivity: Mouse, Isotype: IgG1, kappa, Conjugate: Purified, Formulation: Phosphate-buffered aqueous solution,