Anti-IFNG Rat Monoclonal Antibody [clone: R4-6A2]

Supplier: BIOGEMS INTERNATIONAL INC.
80822-25-500
75843-500EA 684.55 USD
75843-500
Anti-IFNG Rat Monoclonal Antibody [clone: R4-6A2]
Antibodies
The R4-6A2 monoclonal antibody specifically reacts with mouse interferon gamma (IFN-g), an acid-labile interferon produced by CD4 and CD8 T lymphocytes as well as activated NK cells. IFN gamma receptors are present in most immune cells, which respond to its signaling by increasing the surface expression of class I MHC proteins. This promotes the presentation of antigen to T-helper (CD4+) cells. Additionally, it stimulates a number of lymphoid cell functions, including the anti-microbial and anti-tumor responses of macrophages, NK cells, and neutrophils.

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: R4-6A2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR