Anti-IFNG Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: XMG1.2]

Supplier: BIOGEMS INTERNATIONAL INC.
80812-60-25
75843-484EA 83.87 USD
75843-484
Anti-IFNG Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: XMG1.2]
Antibodies
The XMG1.2 is a neutralizing antibody that binds with the mouse Interferon-gamma (IFN-γ) protein, a 15 -17 kDa cytokine with significant antibacterial, antiviral, and antitumoral properties. When secreted by natural killer cells and by natural killer T lymphocytes, it regulates the immune response and supports adaptive immunity when produced by Th1 or CD8+ T lymphocytes. IFN-γ plays an important role in the activation, the growth, and the differentiation of the macrophages, B and T lymphocytes, and natural killer cells. It interacts synergically with other cytokines, such as TNF-α, to inhibit proliferation of normal and transformed cells. IFN- γ is the primary cytokine that defines Th-1 cells.The biological activity of IFN-γ is not affected by glycosylation.

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: XMG1.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR