Anti-IFNG Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 4S.B3]

Supplier: Biogems
80811-50-25
75843-450EA 224.07 USD
75843-450
Anti-IFNG Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 4S.B3]
Antibodies
The 4S.B3 is a neutralizing antibody that binds with the human Interferon-gamma (IFN-γ) protein, a 15 -17 kDa cytokine with significant antibacterial, antiviral, and antitumoral properties. When secreted by natural killer cells and by natural killer T lymphocytes, it regulates the immune response and supports adaptive immunity when produced by Th1 or CD8+ T lymphocytes. IFN-γ plays an important role in the activation, the growth, and the differentiation of the macrophages, B and T lymphocytes, and natural killer cells. It interacts synergically with other cytokines, such as TNF-α, to inhibit proliferation of normal and transformed cells. IFN- γ is the primary cytokine that defines Th-1 cells.The biological activity of IFN-γ is not affected by glycosylation

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: 4S.B3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR