Anti-MCP-1 Armenian Hamster Monoclonal Antibody [clone: 2H5]

Supplier: Biogems
80111-25-50
75843-444EA 217.47 USD
75843-444
Anti-MCP-1 Armenian Hamster Monoclonal Antibody [clone: 2H5]
Antibodies
The 2H5 monoclonal antibody specifically reacts with mouse, rat, human MCP-1 (Monocyte Chemoattractant Protein-1), also known as CCL2 and MCAF (Monocyte Chemotactic and Activating Factor). MCP-1 is a C-C (beta) chemokine that regulates cytokine production in monocytes and the expression of adhesion molecules. The 2h5 antibody can neutralize the bioactivity of recombinant or natural MCP-1.

Type: Primary
Antigen: MCP-1
Clonality: Monoclonal
Clone: 2H5
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR