Anti-TLR9 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M9.D6]

Supplier: BIOGEMS INTERNATIONAL INC.
31912-50-25
75843-288EA 294.1 USD
75843-288
Anti-TLR9 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M9.D6]
Antibodies
The M9.D6 monoclonal antibody specifically reacts with mouse CD289 also known as toll-like receptor 9 (TLR9). CD289 is involved in the activation of innate immunity, in acquired immune responses, and autoimmune diseases. It is involved in the immune system’s response to unmethylated CpG dinucleotide sequences such as those found in bacterial, viral, or synthetic DNA. CD289 assists in pathogen recognition and is expressed by subtypes of dendritic cells and B cells.

Type: Primary
Antigen: TLR9
Clonality: Monoclonal
Clone: M9.D6
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR