Anti-TLR2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TL2.1]

Supplier: BIOGEMS INTERNATIONAL INC.
24911-50-100
75843-232EA 576.34 USD
75843-232
Anti-TLR2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TL2.1]
Antibodies
The TL2.1 monoclonal antibody specifically reacts with human CD282, the Toll-like receptor 2 (TLR2). It is a type I transmembrane signaling receptor containing IL-1 receptor like intracellular domain and leucine-rich repeats (LRR) in the extracellular domain. CD282 is expressed on peripheral blood monocytes and is involved distinguishing bacterial lipoproteins. The TL2.1 antibody blocks the receptor and immunoprecipitates TLR2 from human mammary epithelial cells (HMEC) and peripheral blood mononuclear cells (PBMC).

Type: Primary
Antigen: TLR2
Clonality: Monoclonal
Clone: TL2.1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR