Anti-CD272 Mouse Monoclonal Antibody (Biotin) [clone: MIH26]

Supplier: Biogems
24811-30-25
75843-226EA 274.87 USD
75843-226
Anti-CD272 Mouse Monoclonal Antibody (Biotin) [clone: MIH26]
Antibodies
The MIH26 monoclonal antibody specifically reacts with human CD272, the human and B and T lymphocyte attenuator (BTLA). It is expressed on macrophage, dendritic, NK, NKT, B, and T cells. CD272 (BTLA) is an inhibitory receptor that inhibits T cell proliferation after binding with its ligand, the herpes virus entry mediator (HVEM). It is in the CD28-like family of receptors and is structurally similar to CTLA-4 and PD-1.

Type: Primary
Antigen: CD272
Clonality: Monoclonal
Clone: MIH26
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR