Anti-OX40L Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: RM134L]

Supplier: Biogems
24611-80-25
75843-218EA 103.23 USD
75843-218
Anti-OX40L Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: RM134L]
Antibodies
The RM134L monoclonal antibody specifically reacts with mouse CD252, a TNF/NGF superfamily member present on activated B lymphocytes and antigen-presenting cells. On activated B cells it enhances immunoglobulin secretion and cell proliferation. CD252 is also known as the OX-40 ligand and interacts with the OX-40 antigen found on the surface of activated T cells. The RML134L is reported to block the costimulatory activity of OX-40L.

Type: Primary
Antigen: OX40L
Clonality: Monoclonal
Clone: RM134L
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR