Anti-NOK-1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: NOK-1]

Supplier: BIOGEMS INTERNATIONAL INC.
20811-80-100
75843-190EA 322.61 USD
75843-190
Anti-NOK-1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: NOK-1]
Antibodies
The NOK-1 monoclonal antibody specifically reacts with human CD178, which is the CD95 or Fas ligand. CD178 is a TNF superfamily type II transmembrane glycoprotein expressed by activated T and NK cells and is involved in Fas-mediated apoptosis of lymphocytes. CD178 is also expressed by monocytes, neutrophils, granulocytes and the parenchymal cells of the retina and cornea. The NOK-1 antibody has been reported to bind to COOH-terminus of the Fas ligand in the region associated with Fas binding.

Type: Primary
Antigen: NOK-1
Clonality: Monoclonal
Clone: NOK-1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR