CD138 (Syndecan-1) MAB 25 tests
Supplier: Biogems
CD138 (Syndecan-1), monoclonal Antibody, Clone: DL-101, Host: Mouse, Species reactivity: Human, Isotype: IgG1, kappa, Conjugate: PE, Formulation: Phosphate-buffered aqueous solution,