Anti-CD197 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3D12]

Supplier: Biogems
20011-60-25
75843-174EA 243.02 USD
75843-174
Anti-CD197 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3D12]
Antibodies
The 3D12 monoclonal antibody specifically reacts with human CD197 (CCR7 or EBI-1), a seven-transmembrane G-protein-coupled. It is the receptor for the secondary lymphoid-tissue chemokine (SLC or 6CKine), CCL19, and CCL21. CD197 is expressed on subsets of B and T cells, and mature dendritic cells. The monoclonal 3D12 antibody recognizes the N-terminus of the receptor.

We recommend that this antibody be incubated for at least 45 minutes at 4°C for optimal staining.

Type: Primary
Antigen: CD197
Clonality: Monoclonal
Clone: 3D12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR