Anti-CD135 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: A2F10]

Supplier: BIOGEMS INTERNATIONAL INC.
17412-60-100
75843-148EA 460.49 USD
75843-148
Anti-CD135 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: A2F10]
Antibodies
The A2F10 monoclonal antibody specifically reacts with mouse CD135, a tyrosine kinase class III receptor. CD135, also known as Flt3, Ly-72, and Flk-2, is the receptor for the FLT3 ligand (FLT3L) cytokine and is expressed on many hematopoietic progenitor cells. Signaling through CD135 plays a role in cell survival, proliferation, and differentiation. The A2F10 antibody is reported to inhibit the binding of FLT3L to CD135.

Type: Primary
Antigen: CD135
Clonality: Monoclonal
Clone: A2F10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR