Anti-CD144 (VE-Cadherin) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 16B1]

Supplier: BIOGEMS INTERNATIONAL INC.
16511-60-100
75843-146EA 720.1 USD
75843-146
Anti-CD144 (VE-Cadherin) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 16B1]
Antibodies
The 16B1 monoclonal antibody specifically reacts with human CD144, the 140 kDA molecule called vascular endothelial (VE-cadherin) or cadherin 5. CD144 is an endothelial specific calcium-dependent adhesion molecule involved in cell contact-dependent growth inhibition, migration, survival, adhesion. It is concentrated at the intercellular boundaries of endothelial cells and essential in maintaining cell layer integrity.

Type: Primary
Antigen: CD144 (VE-Cadherin)
Clonality: Monoclonal
Clone: 16B1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR