Anti-CD140a Rat Monoclonal Antibody (Biotin) [clone: APA5]

Supplier: Biogems
12512-30-100
75843-122EA 416.98 USD
75843-122
Anti-CD140a Rat Monoclonal Antibody (Biotin) [clone: APA5]
Antibodies
The APA5 monoclonal antibody specifically binds to the mouse CD140a molecule, otherwise known as the Platelet-Derived Growth Factor (PDGF) Receptor alpha chain. CD140a is a receptor tyrosine kinase expressed on embryonic tissues and adult mesenchymal-derived cells. It binds to PDGF A and B chains and is reported to be involved with wound healing and exhibits chemo-attractant properties. The APA5 antibody blocks the binding of PDGF-AA to the PDGF receptor.

Type: Primary
Antigen: CD140a
Clonality: Monoclonal
Clone: APA5
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR