Anti-CD69 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: H1.2F3]

Supplier: BIOGEMS INTERNATIONAL INC.
11712-60-200
75843-112EA 627.43 USD
75843-112
Anti-CD69 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: H1.2F3]
Antibodies
The H1.2F3 monoclonal antibody specifically reacts with human CD69, the 27-33 kDA type II transmembrane protein also known as the very early activation antigen (VEA) or the activation inducer molecule (AIM). It is expressed as a disulfide-linked dimer on B cells, T cells, NK cells, platelets, eosinophils, and neutrophils. It increases in expression upon cell activation and seems to serve a role as a signaling receptor.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR