Anti-CD8a Mouse Monoclonal Antibody (APC (Allophycocyanin)-Cy7®) [clone: RPA-T8]

Supplier: Biogems
10131-87-25
75843-076EA 148.4 USD
75843-076
Anti-CD8a Mouse Monoclonal Antibody (APC (Allophycocyanin)-Cy7®) [clone: RPA-T8]
Antibodies
The RPA-T8 monoclonal antibody specifically reacts with the human CD8a molecule, a 32 kDa cell surface receptor expressed either as a heterodimer (CD8 α/β) or as a homodimer (CD8 α/α) on the majority of thymocytes, a subpopulation of mature T cells, and natural killer cells. CD8 interacts with the major histocompatibility complex class I (MHC class I) molecules on antigen-presenting cells or epithelial cells. The RPA-T8 antibody reacts with 13-48% of peripheral lymphocytes, 80% of thymocytes, and a subset of natural killer cells. RPA-T8, OKT8, and HIT8a antibodies do not compete with each other for binding to peripheral leukocytes, meaning that that they do not recognize the same epitope or block each other by steric hindrance.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: RPA-T8
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR