Anti-CD5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UCHT2]

Supplier: BIOGEMS INTERNATIONAL INC.
09111-80-100
75843-038EA 283.89 USD
75843-038
Anti-CD5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UCHT2]
Antibodies
The UCHT2 monoclonal antibody specifically reacts with human CD5, a 67 kda type 1 transmembrane glycoprotein. CD5 is expressed on mature T cells, a subset of B cells, and peripheral blood dendritic cells. B cells that are CD5+ produce mostly IgM polyreactive antibodies. Its ligand is CD72, which is involved in T, B cell proliferation and interaction. The UCT2 antibody also recognizes non-human primate CD5.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR