Anti-CD86 (B7-2) (PO3.1) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: PO3.1]

Supplier: Biogems
08922-60-100
75843-026EA 219.36 USD
75843-026
Anti-CD86 (B7-2) (PO3.1) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: PO3.1]
Antibodies
The PO3.1 monoclonal antibody specifically reacts with CD86, also known as B7-2, a surface 80 kDa receptor and member of the B7 family. CD86 is a costimulatory molecule, expressed on B and T cells, dendritic cells, astrocytes, macrophages, Langerhans cells, and at a low level in newly explanted B and T lymphocytes. CD86 expression on B lymphocytes is upregulated by B-Cell Receptor complex, CD40 and certain cytokine receptors. CD80 (B7-1) and CD 86 (B7-2) are receptors for CD28 and CTLA-4, on the surface of T lymphocytes, enhancing the interactions between B and T cells.

Type: Primary
Antigen: CD86 (B7-2) (PO3.1)
Clonality: Monoclonal
Clone: PO3.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR