Anti-CD86 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: GL-1 (GL1)]

Supplier: BIOGEMS INTERNATIONAL INC.
08912-50-500
75843-000EA 270.98 USD
75843-000
Anti-CD86 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: GL-1 (GL1)]
Antibodies
The GL-1 monoclonal antibody reacts specifically with the mouse B7-2 (also known as CD86), a costimulatory molecule expressed by B and T lymphocytes, macrophages (thioglycollate-induced from peritoneum), astrocytes and dendritic cells. The memory CD4+ T lymphocytes express CD86 (as mRNA and protein). CD86, and the B7-1 (CD 80) molecule, are ligands for CD152 and CD28, and influence the costimulatory interactions between lymphocytes B and T. B7-2 is also involved in the mouse natural killer cell-mediated cytotoxicity. The GL-1 antibody blocks the mixed lymphocyte reaction (MLR) and inhibits the T-cells stimulation by antigen-presenting cells. Mixtures of anti-B7-1 antibody and GL-1 were reported to inhibit the interaction between CD125 and its ligand with the in vivo priming of cytotoxic T lymphocytes.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: GL-1 (GL1)
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR