Anti-CD16/32 Mab 100ug
Supplier: Biogems
Anti-CD16/32, Monoclonal antibody, Clone: 2.4G2, Host: Rat, Species: Mouse, Isotype: IgG2b, Conjugate: Safire purified, Formulation: Phosphate-buffered aqueous solution, less than or equal to 0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2. Fc, Size: 100 ug