Anti-CD16 /CD32 Rat Monoclonal Antibody [clone: 2.4G2]

Supplier: Biogems
08212-20-1000
75842-964EA 477.44 USD
75842-964
Anti-CD16 /CD32 Rat Monoclonal Antibody [clone: 2.4G2]
Antibodies
The 2.4G2 monoclonal antibody specifically reacts with an epitope on the extracellular domain of the mouse CD16 (Fc γ III) and CD 32 (Fc γ II). CD16 and CD32 are low affinity receptors for the IgG Fc domain and are expressed by B lymphocytes, NK cells, kupffer cells, mast cells, monocytes, macrophages, granulocytes, immature thymocytes, neutrophils, and some activated mature T cells.The 2.4G2 antibody blocks the binding of immunoglobulins to CD16 and CD32, and possibly to Fc γ I receptor.

Type: Primary
Antigen: CD16 /CD32
Clonality: Monoclonal
Clone: 2.4G2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR