Anti-CD16 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CB16]

Supplier: BIOGEMS INTERNATIONAL INC.
08211-50-25
75842-960EA 198.19 USD
75842-960
Anti-CD16 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CB16]
Antibodies
The CB16 monoclonal antibody specifically reacts with human CD16, the low affinity IgG receptor III (FC gamma RIII). CD16 is expressed on granulocytes, monocytes, macrophages, and NK cells and plays a role in NK activation and signal transduction. CD16 is expressed as either CD16a or CD16b. CD16a is a polypeptide-anchored transmembrane protein and CD16b is a glycosylphosphatidylinositol (GPI)-anchored protein that is expressed exclusively on neutrophils.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: CB16
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR