Anti-CD16 Mouse Monoclonal Antibody [clone: CB16]

Supplier: BIOGEMS INTERNATIONAL INC.
08211-25-500
75842-956EA 889.14 USD
75842-956
Anti-CD16 Mouse Monoclonal Antibody [clone: CB16]
Antibodies
The CB15 monoclonal antibody specifically binds to human CD16, a low affinity Fc receptor. It is expressed on NK, neutrophils, macrophages, monocytes, and placental trophoblasts. CD16 is expressed in CD16a and CD16b forms. CD16a is a polyeptide anchored transmembrane protein and CD16b is a monomeric glycosylphosphatidylinositol (GPI)-anchored protein found in neutrophils. CD16 is reported to be involved in NK cell activation and signal transduction.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: CB16
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR