Anti-CD44 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy5®) [clone: IM7]

Supplier: Biogems
06511-65-25
75842-938EA 90.32 USD
75842-938
Anti-CD44 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy5®) [clone: IM7]
Antibodies
The IM7 monoclonal antibody specifically reacts with all the isoforms and both alloantigens of the CD44 glycoprotein (Pgp-1, Ly-24). CD44 is expressed on hematopoietic and non-hematopoietic cells, bone marrow myeloid cells, memory T lymphocytes, periphery activated B cells, CD4+ T lymphocytes, and CD8+ T lymphocytes. The periphery B and T lymphocytes upregulate the expression of CD44. CD44 binds to hyaluronan molecules, acting as an adhesion molecule.The IM7 antibody inhibits collagen-induced arthritis in DBA/1 mice, prevents central nervous system inflammation associated with experimental autoimmune encephalomyelitis, but exacerbates the experimental autoimmune thyroiditis in CBA/J mice. The IM7 antibody also cross-reacts with dog, human, pig, horse, cat, and cow leukocytes.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: PE (Phycoerythrin)-Cy5®
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR