Anti-CD44 Mab 100ug
Supplier: Biogems
Anti-CD44, Monoclonal antibody, Clone: IM7, Host: Rat, Species: HUman, Mouse, Isotype: IgG2b, kappa, Conjugate: PE-Cy5, Formulation: Phosphate-buffered aqueous solution, less than or equal to 0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2. Fc, Size: 100 ug