Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: OKT3]

Supplier: Biogems
05121-77-25
75842-918EA 122.58 USD
75842-918
Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: OKT3]
Antibodies
The OKT3 monoclonal antibody specifically reacts with the ε chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains γ, δ, and ε chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction.The OKT3 antibody is an immunosuppressive, which has proven to be an effective therapeutic agent in liver, heart, and renal allograft rejection.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR