Anti-CD3 Mab 25tests
Supplier: Biogems
Anti-CD3, Monoclonal antibody, Clone: OKT3, Host: Mouse, Species: Human, Isotype: IgG2a, kappa, Conjugate: BG Violet 450, Formulation: Phosphate-buffered aqueous solution, less than or equal to 0.09% Sodium azide, may contain carrier protein/stabilizer, Fc, Size: 25 tests